Assay Details
| Gene Symbol: PRDM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451241.1 | XP_015306727.1 | 8 - 9 | 1030 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM9| Gene Symbol: PRDM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451241.1 | XP_015306727.1 | 8 - 9 | 1030 | 120 | |
Set Membership: |
|