Assay Details
| Gene Symbol: FOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568193.2 | XP_005568250.1 | 1 - 2 | 386 | 63 | |
| XM_005568194.2 | XP_005568251.1 | 2 - 3 | 316 | 63 | ||
More Information
Set Membership: |
|

FOXA2
Genomic Map