Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546108.2 | XP_005546165.1 | 2 - 3 | 493 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005546108.2 transcripts.
Set Membership: |
|

GPR87
Genomic Map