Assay Details
| Gene Symbol: SLC25A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558047.2 | XP_005558104.1 | 1 | 456 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005558047.2 transcripts.
Set Membership: |
|

SLC25A2
Genomic Map