Assay Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546491.2 | XP_005546548.1 | 2 | 4482 | 117 | |
| XM_005546492.2 | XP_005546549.1 | 3 | 4662 | 117 | ||
| XM_005546494.2 | XP_005546551.1 | 2 | 3767 | 117 | ||
More Information
Set Membership: |
|

B3GNT5
Genomic Map