Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563739.2 | XP_005563796.1 | 4 - 5 | 1910 | 95 | |
| XM_005563742.2 | XP_005563799.1 | 3 - 4 | 1745 | 95 | ||
| XM_015455048.1 | XP_015310534.1 | 3 - 4 | 1667 | 95 | ||
| XM_015455049.1 | XP_015310535.1 | 5 - 6 | 2006 | 95 | ||
More Information
Set Membership: |
|

TP53INP1
Genomic Map