Assay Details
| Gene Symbol: CHST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559187.2 | XP_005559244.1 | 1 | 214 | 167 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005559187.2 transcripts.
Set Membership: |
|

CHST14
Genomic Map