Assay Details
| Gene Symbol: LIPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575056.2 | XP_005575113.1 | 2 | 769 | 164 | |
| XM_005575057.2 | XP_005575114.1 | 2 | 909 | 164 | ||
More Information
Set Membership: |
|

LIPT1
Genomic Map