Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571004.2 | XP_005571061.1 | 2 - 3 | 2842 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005571004.2 transcripts.
Set Membership: |
|

SP7
Genomic Map