Assay Details
| Gene Symbol: P2RX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582526.2 | XP_005582583.1 | 4 - 5 | 824 | 119 | |
| XM_005582527.2 | XP_005582584.1 | 4 - 5 | 824 | 119 | ||
| XM_015437456.1 | XP_015292942.1 | 4 - 5 | 824 | 119 | ||
| XM_015437457.1 | XP_015292943.1 | 3 - 4 | 806 | 119 | ||
| XM_015437458.1 | XP_015292944.1 | 3 - 4 | 845 | 119 | ||
More Information
Set Membership: |
|

P2RX5
Genomic Map