Assay Details
| Gene Symbol: ARSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557233.2 | XP_005557290.1 | 1 - 2 | 1577 | 87 | |
| XM_005557234.2 | XP_005557291.1 | 1 - 2 | 1479 | 87 | ||
More Information
Set Membership: |
|

ARSB
Genomic Map