Assay Details
| Gene Symbol: CLCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593575.2 | XP_005593632.1 | 9 - 10 | 1313 | 97 | |
| XM_005593576.2 | XP_005593633.1 | 9 - 10 | 1218 | 97 | ||
| XM_005593577.2 | XP_005593634.1 | 10 - 11 | 1468 | 97 | ||
| XM_005593579.2 | XP_005593636.1 | 6 - 7 | 3142 | 97 | ||
| XM_015443504.1 | XP_015298990.1 | 9 - 10 | 1578 | 97 | ||
| XM_015443505.1 | XP_015298991.1 | 7 - 8 | 1026 | 97 | ||
| XM_015443506.1 | XP_015298992.1 | 6 - 7 | 794 | 97 | ||
More Information
Set Membership: |
|

CLCN5
Genomic Map