Assay Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565882.2 | XP_005565939.1 | 8 - 9 | 2076 | 73 | |
| XM_015456364.1 | XP_015311850.1 | 7 - 8 | 920 | 73 | ||
| GenBank mRNA | AB169328.1 | - | 7 - 8 | 925 | 73 | |
More Information
Set Membership: |
|

PTEN
Genomic Map