Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555691.2 | XP_005555748.1 | 2 - 3 | 1063 | 142 | |
| XM_005555692.2 | XP_005555749.1 | 4 - 5 | 1814 | 142 | ||
| XM_005555693.2 | XP_005555750.1 | 3 - 4 | 1668 | 142 | ||
More Information
Set Membership: |
|

PITX2
Genomic Map