Assay Details
| Gene Symbol: HNF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569073.2 | XP_005569130.1 | 6 - 7 | 844 | 91 | |
| XM_005569074.2 | XP_005569131.1 | 6 - 7 | 844 | 91 | ||
| XM_005569075.2 | XP_005569132.1 | 6 - 7 | 718 | 91 | ||
| XM_015429862.1 | XP_015285348.1 | 6 - 7 | 718 | 91 | ||
More Information
Set Membership: |
|

HNF4A
Genomic Map