Assay Details
| Gene Symbol: HEXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284738.1 | NP_001271667.1 | 6 - 7 | 656 | 94 | |
| XM_015452428.1 | XP_015307914.1 | 6 - 7 | 1176 | 94 | ||
| GenBank mRNA | AB169218.1 | - | 6 - 7 | 691 | 94 | |
| AB170139.1 | - | 6 - 7 | 656 | 94 | ||
More Information
Set Membership: |
|

HEXA
Genomic Map