Assay Details
| Gene Symbol: IL1RN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283539.1 | NP_001270468.1 | 2 - 3 | 204 | 125 | |
| GenBank mRNA | AY182232.1 | - | 2 - 3 | 208 | 125 | |
More Information
Set Membership: |
|

IL1RN
Genomic Map