Assay Details
| Gene Symbol: CCKAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554633.2 | XP_005554690.1 | 2 - 3 | 429 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCKAR| Gene Symbol: CCKAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554633.2 | XP_005554690.1 | 2 - 3 | 429 | 134 | |
Set Membership: |
|