Assay Details
| Gene Symbol: GRM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550666.2 | XP_005550723.1 | 3 - 4 | 1532 | 132 | |
| XM_005550668.2 | XP_005550725.1 | 3 - 4 | 1532 | 132 | ||
| XM_005550669.2 | XP_005550726.1 | 3 - 4 | 1356 | 132 | ||
More Information
Set Membership: |
|

GRM8
Genomic Map