Assay Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553929.2 | XP_005553986.1 | 5 - 6 | 683 | 145 | |
| XM_005553930.2 | XP_005553987.1 | 4 - 5 | 1008 | 145 | ||
| XM_015449854.1 | XP_015305340.1 | 5 - 6 | 680 | 145 | ||
More Information
Set Membership: |
|

PRL
Genomic Map