Assay Details
Gene Symbol: BICD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582257.2 | XP_005582314.1 | 2 - 3 | 591 | 86 | |
XM_005582258.2 | XP_005582315.1 | 2 - 3 | 591 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
