Assay Details
| Gene Symbol: DUSP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565409.2 | XP_005565466.1 | 3 - 4 | 609 | 153 | |
| XM_005565412.2 | XP_005565469.1 | 2 - 3 | 410 | 153 | ||
| XM_015456138.1 | XP_015311624.1 | 4 - 5 | 818 | 153 | ||
More Information
Set Membership: |
|

DUSP13
Genomic Map