Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564526.1 | XP_005564583.1 | 1 - 2 | 369 | 135 | |
| GenBank mRNA | AB170848.1 | - | 1 - 2 | 286 | 135 | |
| AB173024.1 | - | 1 - 2 | 319 | 118 | ||
More Information
Set Membership: |
|

KLF6
Genomic Map