Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550933.2 | XP_005550990.1 | 3 - 4 | 720 | 114 | |
XM_015448138.1 | XP_015303624.1 | 3 - 4 | 705 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
