Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572809.2 | XP_005572866.1 | 3 - 4 | 409 | 94 | |
| XM_005572810.2 | XP_005572867.1 | 3 - 4 | 244 | 94 | ||
More Information
Set Membership: |
|

FAM168B
Genomic Map