Assay Details
| Gene Symbol: PLPPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542623.2 | XP_005542680.1 | 1 - 2 | 454 | 76 | |
| XM_005542624.2 | XP_005542681.1 | 1 - 2 | 454 | 76 | ||
| XM_005542625.2 | XP_005542682.1 | 2 - 3 | 3696 | 76 | ||
| XM_015432347.1 | XP_015287833.1 | 1 - 2 | 545 | 76 | ||
| XM_015432350.1 | XP_015287836.1 | 1 - 2 | 545 | 76 | ||
More Information
Set Membership: |
|

PLPPR5
Genomic Map