Assay Details
| Gene Symbol: TSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553084.2 | XP_005553141.1 | 3 - 4 | 647 | 108 | |
| XM_015449444.1 | XP_015304930.1 | 3 - 4 | 379 | 108 | ||
More Information
Set Membership: |
|

TSPO2
Genomic Map