Assay Details
| Gene Symbol: C1H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541697.2 | XP_005541754.1 | 2 - 3 | 392 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005541697.2 transcripts.
Set Membership: |
|

C1H1orf189
Genomic Map