Assay Details
| Gene Symbol: MYBPHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015431701.1 | XP_015287187.1 | 1 - 2 | 5121 | 97 | |
| XM_015431712.1 | XP_015287198.1 | 1 - 2 | 5121 | 97 | ||
| XM_015431720.1 | XP_015287206.1 | 1 - 2 | 5121 | 97 | ||
| XM_015431728.1 | XP_015287214.1 | 1 - 2 | 5121 | 97 | ||
More Information
Set Membership: |
|

MYBPHL
Genomic Map