Assay Details
| Gene Symbol: CNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585932.2 | XP_005585989.1 | 3 - 4 | 695 | 98 | |
| XM_005585933.2 | XP_005585990.1 | 3 - 4 | 699 | 98 | ||
More Information
Set Membership: |
|

CNMD
Genomic Map