Assay Details
Gene Symbol: C2H3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547089.2 | XP_005547146.1 | 1 - 2 | 1493 | 118 | |
XM_005547090.2 | XP_005547147.1 | 2 - 3 | 486 | 118 | ||
XM_015445905.1 | XP_015301391.1 | 2 - 3 | 553 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
