Assay Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539636.2 | XP_005539693.2 | 1 - 2 | 1922 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005539636.2 transcripts.
Set Membership: |
|

ZBTB18
Genomic Map