Assay Details
| Gene Symbol: C2CD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559703.2 | XP_005559760.1 | 2 | 967 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005559703.2 transcripts.
Set Membership: |
|

C2CD4A
Genomic Map