Assay Details
| Gene Symbol: PPP1R42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319560.1 | NP_001306489.1 | 4 - 5 | 523 | 128 | |
| XM_005563457.2 | XP_005563514.1 | 3 - 4 | 480 | 128 | ||
| GenBank mRNA | AB168658.1 | - | 4 - 5 | 523 | 128 | |
More Information
Set Membership: |
|

PPP1R42
Genomic Map