Assay Details
| Gene Symbol: SLC35A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593499.2 | XP_005593556.1 | 2 - 3 | 1080 | 79 | |
| XM_015443424.1 | XP_015298910.1 | 2 - 3 | 1082 | 79 | ||
| XM_015443425.1 | XP_015298911.1 | 3 - 4 | 1178 | 79 | ||
| GenBank mRNA | AB169772.1 | - | 2 - 3 | 286 | 79 | |
| AB170449.1 | - | 2 - 3 | 287 | 79 | ||
More Information
Set Membership: |
|

SLC35A2
Genomic Map