Assay Details
| Gene Symbol: RDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285270.1 | NP_001272199.1 | 3 - 4 | 442 | 107 | |
| XM_005583490.2 | XP_005583547.1 | 3 - 4 | 454 | 107 | ||
| XM_015437861.1 | XP_015293347.1 | 2 - 3 | 273 | 107 | ||
| GenBank mRNA | AB168602.1 | - | 3 - 4 | 442 | 107 | |
More Information
Set Membership: |
|

RDM1
Genomic Map