Assay Details
| Gene Symbol: PRSS53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591712.2 | XP_005591769.1 | 9 - 10 | 2284 | 112 | |
| XM_005591713.2 | XP_005591770.1 | 9 - 10 | 2284 | 112 | ||
| XM_015442442.1 | XP_015297928.1 | 5 - 6 | 1526 | 112 | ||
More Information
Set Membership: |
|

PRSS53
Genomic Map