Assay Details
| Gene Symbol: AP1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574437.2 | XP_005574494.1 | 4 - 5 | 713 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AP1S3| Gene Symbol: AP1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574437.2 | XP_005574494.1 | 4 - 5 | 713 | 117 | |
Set Membership: |
|