Assay Details
| Gene Symbol: ZMYM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319638.2 | NP_001306567.2 | 2 - 3 | 88 | 126 | |
| XM_005585458.1 | XP_005585515.1 | 2 - 3 | 158 | 126 | ||
| XM_015439038.1 | XP_015294524.1 | 2 - 3 | 158 | 126 | ||
| XM_015439039.1 | XP_015294525.1 | 2 - 3 | 81 | 126 | ||
| XM_015439040.1 | XP_015294526.1 | 2 - 3 | 227 | 126 | ||
| XM_015439042.1 | XP_015294528.1 | 2 - 3 | 158 | 126 | ||
| GenBank mRNA | DC635798.1 | - | 2 - 3 | 88 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001319638.2, XM_005585458.1, XM_015439038.1, XM_015439039.1, XM_015439040.1, XM_015439042.1 transcripts.
Set Membership: |
|

ZMYM5
Genomic Map