Assay Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577005.2 | XP_005577062.1 | 4 - 5 | 920 | 139 | |
| XM_015434340.1 | XP_015289826.1 | 4 - 5 | 920 | 139 | ||
| XM_015434341.1 | XP_015289827.1 | 4 - 5 | 920 | 139 | ||
| XM_015434342.1 | XP_015289828.1 | 4 - 5 | 920 | 139 | ||
More Information
Set Membership: |
|

TESMIN
Genomic Map