Assay Details
| Gene Symbol: SYCP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449900.1 | XP_015305386.1 | 7 - 8 | 460 | 96 | |
| XM_015449901.1 | XP_015305387.1 | 7 - 8 | 544 | 96 | ||
| XM_015449902.1 | XP_015305388.1 | 10 - 11 | 860 | 96 | ||
More Information
Set Membership: |
|

SYCP2L
Genomic Map