Assay Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592215.2 | XP_005592272.1 | 5 - 6 | 2292 | 147 | |
XM_005592216.2 | XP_005592273.1 | 5 - 6 | 1875 | 147 | ||
XM_005592217.2 | XP_005592274.1 | 5 - 6 | 1722 | 147 | ||
XM_005592218.2 | XP_005592275.1 | 5 - 6 | 748 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
