Assay Details
| Gene Symbol: SLC2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582724.2 | XP_005582781.1 | 8 - 9 | 1278 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A4| Gene Symbol: SLC2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582724.2 | XP_005582781.1 | 8 - 9 | 1278 | 151 | |
Set Membership: |
|