Assay Details
| Gene Symbol: TCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577747.2 | XP_005577804.1 | 5 - 6 | 954 | 141 | |
| XM_015434664.1 | XP_015290150.1 | 5 - 6 | 961 | 141 | ||
More Information
Set Membership: |
|

TCN1
Genomic Map