Assay Details
| Gene Symbol: LOC102135442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575029.2 | XP_005575086.1 | 1 - 2 | 252 | 112 | |
| XM_005575030.2 | XP_005575087.1 | 1 - 2 | 137 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005575029.2, XM_005575030.2 transcripts.
Set Membership: |
|

LOC102135442
Genomic Map