Assay Details
| Gene Symbol: LOC102135442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575029.2 | XP_005575086.1 | 6 - 7 | 1138 | 137 | |
| XM_005575030.2 | XP_005575087.1 | 6 - 7 | 961 | 137 | ||
More Information
Set Membership: |
|

LOC102135442
Genomic Map