Assay Details
| Gene Symbol: ALDH5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553921.2 | XP_005553978.1 | 7 - 8 | 1400 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALDH5A1| Gene Symbol: ALDH5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553921.2 | XP_005553978.1 | 7 - 8 | 1400 | 103 | |
Set Membership: |
|