Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569218.2 | XP_005569275.1 | 7 - 8 | 732 | 166 | |
| XM_005569220.2 | XP_005569277.1 | 5 - 6 | 573 | 166 | ||
| XM_005569221.2 | XP_005569278.1 | 6 - 7 | 667 | 166 | ||
More Information
Set Membership: |
|

CD40
Genomic Map