Assay Details
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585015.2 | XP_005585072.1 | 4 - 5 | 655 | 116 | |
| XM_015438867.1 | XP_015294353.1 | 3 - 4 | 736 | 116 | ||
| XM_015438869.1 | XP_015294355.1 | 3 - 4 | 739 | 116 | ||
More Information
Set Membership: |
|

CDK3
Genomic Map