Assay Details
| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562955.2 | XP_005563012.1 | 3 - 4 | 2908 | 101 | |
| XM_015454636.1 | XP_015310122.1 | 3 - 4 | 2810 | 101 | ||
| GenBank mRNA | AB172049.1 | - | 3 - 4 | 1136 | 101 | |
More Information
Set Membership: |
|

EXTL3
Genomic Map